Läs mer
3M™ Red SMS Based Fabric Category III Protective Coveralls
Handla allt 3M Produkter
:
Size: XX-Large
Specifications
Specifications
| Produkttyp | General Purpose Coveralls |
| Kön | Unisex |
| Disponibel | Single-use |
| Midja stil | Elastic |
| Ankel stil | Elastic |
| Manschettstil | Elastic |
| Hals stil | Hooded |
| Huva typ | 2-panel |
| Sömtyp | Serged |
| Sterilitet | Non-sterile |
| Show More |