Learn More
Eppendorf™ Reference™ 2 EU-IVD Single Channel Variable Volume Pipette
Extraordinary precision and accuracy, a long service lifes and an ergonomic design.
4108.56 SEK
Specifications
Certifications/Compliance | In vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998 |
---|---|
Autoclavable | Autoclavable |
Number of Channels | 1 |
Product Line | Reference™ 2 |
Sterility | Sterile |
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
15163666
|
Eppendorf™
4920000040 |
2 to 20 μL | Yellow |
4108.56 SEK
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15103676
|
Eppendorf™
4920000083 |
100 to 1000 μL | Blue |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15153666
|
Eppendorf™
4920000032 |
2 to 20 μL | Gray |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Extraordinary precision and accuracy, a long service lifes and an ergonomic design.
Specifications
In vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998 | |
1 | |
Sterile | |
4-digit display visible from all angles | |
RFID Tracking |
Autoclavable | |
Reference™ 2 | |
Pipette | |
Forward pipetting, Reverse pipetting, Removal of supernatants, Sample mixing, Phase extraction, Filling of plates, gels, and reaction vessels |