missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ ScienceLine Glass Single Reference Electrode

Especially useful for the separation of combination electrodes into separate measuring and reference electrodes. SI Analytics™ ScienceLine Glass Single Reference Electrode are for special applications and make complex applications even more reliable.

Brand:  SI Analytics™ 1070073

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10023364

  • 1676.68 SEK / Each


Call for latest availability.
+46 (0)31 352 32 30

Add to basket

Description

Description

  • The combination of pure measuring electrodes for i.e. ion-selective measurements or special constructions with one reference electrode
  • Redox titrations with a constant pH value with glass electrodes as reference and redox single sensors as measuring electrode
  • Application of double-electrolyte electrodes for an optimal adaptation of reference electrolyte to the measuring media.
Specifications

Specifications

–5 to 100°C
Platinum Diaphragm
103mm
0 to 14
Reference Electrode
10mm
Glass
Ag/AgCl
Videos