Läs mer
Techne™ Refrigerated Bath
Modell:
RB-12A
RB-5A
Kapacitet (metrisk):
7 l
12 l
Förpackningsstorlek:
Varje
Beskrivning
- Built-in refrigeration for efficient cooling
- Robust stainless steel construction makes for easy cleaning
Specifikationer
Specifikationer
| Kapacitet (metrisk) | 12 l |
| Kylkapacitet | 20°C at 240W, 0°C at 240W, -10° C at 230W |
| Temperaturområde | 35°C to 100°C |
| Funktioner | Circulating bath with integrated refrigeration unit, Temperature range from -35°C to +100°C, Robust stainless-steel construction, Three models of refrigerated bath available: 7, 12, 22 liter,3 year warranty. |
| Modell | RB-12A |
| Garanti | 3 Years |
| Spänning | 230 V |
| Frekvens | 50/60 Hz |