missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ REG3A (Human) Recombinant Protein

Human REG3A full-length ORF ( AAH36776.1, 1 a.a. - 175 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005068-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16162365

  • 4055.00 SEK / 10µg
Estimated Shipment: 14-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MLPPMALPSVSWMLLSCLMLLSQVQGEEPQRELPSARIRCPKGSKAYGSHCYALFLSPKSWTDADLACQKRPSGNLVSVLSGAEGSFVSSLVKSIGNSYSYVWIGLHDPTQGTEPNGEGWEWSSSDVMNYFAWERNPSTISSPGHCASLSRSTAFLRWKDYNCNVRLPYVCKFTD
Specifications

Specifications

AAH36776.1
regenerating islet-derived 3 alpha
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
REG3A
GST
5068
Wheat germ expression system
10 μg
HIP, INGAP, PAP, PAP-H, PAP1, PBCGF, REG-III, REG3
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers