Läs mer
Ismatec™ Reglo Independent Channel Control (ICC) Peristaltic Pumps
Produktkod. 16629762
Flödeshastighet:
24 ml/min
43 ml/min
Kanaler:
3
4
Produktkod. 16629762
Brand: Ismatec™ ISM4306
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Independently control flow and direction of each channel via keypad or PC
  • Flow rate 0.001 to 35 mL/min per channel
  • Flow rate depends on drive rpm and tubing size
  • USB interface for quick connection to a PC
  • Flexibility of bidirectional flow in each channel
Specifikationer
Noggrannhet ±1%
Kanaler 3
Kontrolltyp Digital Variable Speed
Flödeshastighet 43 ml/min
Maximalt tryck 14.5 psi
Reversibel Yes
Antal rullar 6
Spänning 100/240 V