Läs mer
Regorafenib, TRC
Produktkod. 18040910
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18040910 25 mg
18020520 100 mg
18091910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18040910 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR14300025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 755037-03-7
Molekylformel C21 H15 Cl F4 N4 O3
Molekylvikt (g/mol) 482.82
Synonym 2-Pyridinecarboxamide, 4-[4-[[[[4-chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]-3-fluorophenoxy]-N-methyl-, 4-[4-[[[[4-Chloro-3-(trifluoromethyl)phenyl]amino]carbonyl]amino]-3-fluorophenoxy]-N-methyl-2-pyridinecarboxamide, 4-[4-[N'-(4-Chloro-3-trifluoromethylphenyl)ureido]-3-fluorophenoxy]pyridine-2-carboxylic acid methylamide, BAY 73-4506, Regorafenib, Stivarga
IUPAC-namn 4-[4-[[4-chloro-3-(trifluoromethyl)phenyl]carbamoylamino]-3-fluorophenoxy]-N-methylpyridine-2-carboxamide
LEDER CNC(=O)c1cc(Oc2ccc(NC(=O)Nc3ccc(Cl)c(c3)C(F)(F)F)c(F)c2)ccn1
Specifikationer
Kemiskt namn eller material Regorafenib
Kvantitet 25 mg
InChI formel InChI=1S/C21H15ClF4N4O3/c1-27-19(31)18-10-13(6-7-28-18)33-12-3-5-17(16(23)9-12)30-20(32)29-11-2-4-15(22)14(8-11)21(24,25)26/h2-10H,1H3,(H,27,31)(H2,29,30,32)
Formel vikt 482.0769