Learn More
Memmert™ Reinforced Stainless Steel Grid
Additional reinforced stainless steel grid for reinforced chamber option
Brand:  Memmert™ IPP110ecoE29767
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17130105
  • 3325.29 SEK / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • Consider max. loading of chamber; electropolished, max. loading 60 kg
Specifications
Specifications
IPP110eco