Learn More
Memmert™ Reinforced Stainless Steel Grid
Additional reinforced stainless steel grid for reinforced chamber option
Brand:  Memmert™ IPP1060ecoB32550
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 17180045
  • 7715.61 SEK / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Consider max. loading of chamber; electropolished, max. loading 60 kg
Specifications
Specifications
IPP1060eco