Läs mer
rel-trans-Methoprene Epoxide, TRC
Produktkod. 30137223
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137223 100 mg
30003265 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137223 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM260655100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 65277-96-5
Molekylformel C19 H34 O4
Molekylvikt (g/mol) 326.47
Synonym 2-Butenoic acid, 3-[(2R,3R)-3-(6-methoxy-2,6-dimethylheptyl)-2-oxiranyl]-, 1-methylethyl ester, (2E)-rel- (ACI), (2E)-rel-3-[(2R,3R)-3-(6-Methoxy-2,6-dimethylheptyl)-2-oxiranyl]-2-butenoic Acid 1-Methylethyl Ester, (2E)-rel-3-[(2R,3R)-3-(6-Methoxy-2,6-dimethylheptyl)oxiranyl]-2-butenoic Acid 1-Methylethyl Ester, 2-Butenoic acid, 3-[(2R,3R)-3-(6-methoxy-2,6-dimethylheptyl)oxiranyl]-, 1-methylethyl ester, (2E)-rel- (9CI), rel-1-Methylethyl (2E)-3-[(2R,3R)-3-(6-methoxy-2,6-dimethylheptyl)-2-oxiranyl]-2-butenoate (ACI), Methoprene epoxide, rel-trans-Methoprene epoxide, Methoprene trans-epoxide, trans-Methoprene-epoxide
IUPAC-namn propan-2-yl (E)-3-[(2R,3R)-3-(6-methoxy-2,6-dimethylheptyl)oxiran-2-yl]but-2-enoate
LEDER COC(C)(C)CCCC(C)C[C@H]1O[C@@H]1\C(=C\C(=O)OC(C)C)\C
Specifikationer
Kemiskt namn eller material trans-Methoprene-epoxide
Kvantitet 100 mg
InChI formel InChI=1S/C19H34O4/c1-13(2)22-17(20)12-15(4)18-16(23-18)11-14(3)9-8-10-19(5,6)21-7/h12-14,16,18H,8-11H2,1-7H3/b15-12+/t14?,16-,18-/m1/s1
Formel vikt 326.2457