Läs mer
RelA/NFkB p65 Antibody (112A1021), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-56055
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
RelA/NFkB p65 Monoclonal specifically detects RelA/NFkB p65 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifikationer
| RelA/NFkB p65 | |
| Monoclonal | |
| 0.5mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1ul/1 million cells, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Cross reacts with Human, Mouse and Rat. Predicted to react with Cow, Gorilla, New World Monkey and Pig. | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 112A1021 | |
| Biotin | |
| Q969V5 | |
| RELA | |
| This NfkB p65 antibody was raised against a synthetic peptide corresponding to amino acids 526-539 of human NF-kB (p65) protein. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| 5970 | |
| Human, Mouse, Rat | |
| Purified |
Korrigering av produktinnehåll
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering