missing translation for 'onlineSavingsMsg'
Läs mer

RelA/NFkB p65, p Ser276 Antibody, Novus Biologicals™

Produktkod. 18260314 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18260314 0.025 mL
18741104 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18260314 Leverantör Novus Biologicals Leverantörsnummer NB100820860.025ML

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody has been used in 3 publications

RelA/NFkB p65 Polyclonal specifically detects RelA/NFkB p65 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen RelA/NFkB p65 (p Ser276)
Användningsområden Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry
Klassificering Polyclonal
Koncentration 1.0 mg/mL
Konjugera Unconjugated
Utspädning Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200 Immunohistochemistry-Paraffin 1:50-1:100
Formulering phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Genaccessionsnr. Q04206
Gene Alias MGC131774, nf kb p65, NF kB p65 phospho, NFkB p65, NF-kB p65, NFkB p65 phospho, NF-kB p65 phospho, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, phospho NF kB p65, phospho NFkB p65, phospho NF-kB p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Gensymboler RELA
Värd art Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human NF-kB p65 around the phosphorylation site of serine 276 (R-P-SP-D-R).
Molekylvikt av antigen 65 kDa
Reningsmetod Affinity Purified
Kvantitet 0.025 mL
Regulatorisk status RUO
Forskningsdisciplin Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primär eller sekundär Primary
Gen-ID (Entrez) 5970
Testspecificitet NkappaF-B p65 (specific to Phospho-Ser276) detects endogenous levels of NF-B p65 only when phosphorylated at serine 276.
Målarter Mouse, Rat, Human
Innehåll och lagring Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotyp IgG
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.