missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RELT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14532-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is especially abundant in hematologic tissues. It has been shown to activate the NF-kappaB pathway and selectively bind TNF receptor-associated factor 1 (TRAF1). This receptor is capable of stimulating T-cell proliferation in the presence of CD3 signaling, which suggests its regulatory role in immune response. Two alternatively spliced transcript variants of this gene encoding the same protein have been reported.Specifications
RELT | |
Polyclonal | |
Unconjugated | |
RELT | |
RELT, TNFRSF19L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
320100, 361615, 84957 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BX43, Q969Z4 | |
Relt | |
RELT Fusion Protein Ag6032 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |