Learn More
Thermo Scientific™ Remel™ Egg Yolk Tellurite 100%
Remel Egg Yolk Tellurite 100% is used with Baird Parker Agar Base.
288.00 SEK
Specifications
Description Egg Yolk Tellurite 100%
Form Liquid
Product Type Microbiology Media
View More Specs
Products 1
Product Code Brand Packaging Type Quantity Price Quantity & Availability  
Product Code Brand Packaging Type Quantity Price Quantity & Availability  
11983122
View Documents Promotion Details
Thermo Scientific™
R450330
1 Ea. 100 mL
288.00 SEK
100mL
View alternative products
Save Up to 
»
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
Remel Egg Yolk Tellurite 100% is used with Baird Parker Agar Base.
Specifications
Specifications
Egg Yolk Tellurite 100%
Liquid
Microbiology Media