Learn More
Thermo Scientific™ Middlebrook 7H10 Agar (Dehydrated)
Isolate and cultivate Mycobacterium spp. using Middlebrook 7H10 Agar.
Brand:  Thermo Scientific™ R453982
Additional Details : Weight : 0.50000kg
Product Code. 11932652
  • 2065.00 SEK / 500g
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
Description
Description
Isolate and cultivate Mycobacterium spp. using Thermo Scientific™ Remel™ Middlebrook 7H10 Agar (Dehydrated). Dubos and Middlebrook developed media formulations containing oleic acid and albumin which enhanced the growth of tubercle bacilli and protected the organisms against a variety of toxic agents1. In 1958, Middlebrook and Cohn improved the first formulation of oleic acid and albumin agar to obtain 7H10 Agar, which allows faster and more luxuriant growth of Mycobacterium species2.
Isolate and cultivate Mycobacterium spp. using Middlebrook 7H10 Agar.
  • Selective: For Mycobacterium spp from other bacteria due to the presence of malachite-green dye.
  • Allows luxuriant growth of Mycobacterium
This medium is used for qualitative procedures. This medium contains inorganic salts which are essential for the growth of mycobacteria. Malachite-green dye acts as a selective agent which partially inhibits bacteria other than mycobacteria.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
Middlebrook 7H10 Agar
Powder
Agar
500 g
Special Offers
REF R09565 requires addition of OADC enrichment, REF R450600.