Läs mer
Thermo Scientific™ OF Medium w/1% Dextrose
Produktkod. 15127522
Change view
Kvantitet:
20 x 4 mL
Förpackningsstorlek:
Paket med 20
Produktkod. Kvantitet unitSize
15127522 20 x 4 mL Paket med 20
1 options
Produktkod. 15127522 Leverantör Thermo Scientific™ Leverantörsnummer R061918
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Use OF Medium w/1% Dextrose (Hugh-Liefson) to determine oxidative and fermentative metabolism of carbohydrates by gram-negative bacilli.
Determine oxidative and fermentative metabolism of glucose by gram-negative bacilli using Thermo Scientific™ Remel™ OF Medium w/1% Dextrose (Hugh-Liefson). Hugh and Leifson first reported that microorganisms which utilize carbohydrates oxidatively produce only a minimal amount of acid1. Oxidative-Fermentative (OF) Medium was developed with a ratio of peptone to carbohydrate that minimized the formation of alkaline by-products and enhanced acid production by oxidative organisms. OF Medium is especially suitable for determining the carbohydrate metabolic pathway of gram-negative bacilli2.
Typical fermentation media contain approximately 2% peptone and 1% carbohydrate. In media containing high concentrations of peptones, oxidative microorganisms produce alkaline by-products in quantities that neutralize the acid produced from carbohydrat
TRUSTED_SUSTAINABILITY
Specifikationer
Certifikat och efterlevnad Industrial Reference: BAM.
Beskrivning OF Medium w/1% Dextrose (Hugh-Liefson)
Formatera 15mm x 103mm Tube
Produkttyp Microbiology Media
Kvantitet 20 x 4 mL