Learn More
Thermo Scientific™ Remel™ PathoDx™ Extraction Reagent 1
Remel PathoDx and Extraction Reagent 1 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.
Brand: Thermo Scientific™ R62050
Additional Details : Weight : 0.50000kg
Description
Remel PathoDx and Extraction Reagent 1 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.
Specifications
2°C to 8°C | |
Yes | |
Each |
PathoDx Extraction Reagent 1, 7mL/Vial | |
Vial |