missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Vitamin K, Hemin Solution

Remel Vitamin K, Hemin Solution is used in anaerobic media.

Brand:  Thermo Scientific™ R450951

Additional Details : Weight : 0.01000kg

Product Code. 11414119

  • 1730.00 SEK / 200mL
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

Remel Vitamin K, Hemin Solution is used in anaerobic media.

Specifications

Specifications

Vitamin K: 0.5mg/10mL. Hemin: 5mg/10mL.
Liquid
10 mL
Vitamin K, Hemin Solution
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers