Läs mer
Remifentanil Ester Hydrochloride, TRC
Produktkod. 30426516
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30426516 50 mg
30422933 10 mg
30417263 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426516 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR14351550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2732614-50-3
Molekylformel C13H18N2O2 • HCl
Molekylvikt (g/mol) 270.76
Synonym (Phenylamino)methyl Piperidine-4-carboxylate, Methyl 4-anilinopiperidine-4-carboxylate Hydrochloride
IUPAC-namn methyl 4-(phenylamino)piperidine-4-carboxylate hydrochloride
LEDER COC(C1(NC2=CC=CC=C2)CCNCC1)=O.Cl
Specifikationer
Kemiskt namn eller material Remifentanil Ester Hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C13H18N2O2.ClH/c1-17-12(16)13(7-9-14-10-8-13)15-11-5-3-2-4-6-11;/h2-6,14-15H,7-10H2,1H3;1H
Formel vikt 270.12