Läs mer
Repaglinide Acyl-Beta-D-glucuronide, TRC
Produktkod. 30010214
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010214 1 mg
30008033 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010214 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR1445101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1309112-13-7
Molekylformel C33 H44 N2 O10
Molekylvikt (g/mol) 628.71
Synonym Repaglinide Glucuronide
IUPAC-namn (2S,3S,4S,5R)-6-[2-ethoxy-4-[2-[[(1S)-3-methyl-1-(2-piperidin-1-ylphenyl)butyl]amino]-2-oxoethyl]benzoyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
LEDER CCOc1cc(CC(=O)N[C@@H](CC(C)C)c2ccccc2N3CCCCC3)ccc1C(=O)OC4O[C@@H]([C@@H](O)[C@H](O)[C@H]4O)C(=O)O
Specifikationer
Kemiskt namn eller material Repaglinide Acyl-beta-D-glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C33H44N2O10/c1-4-43-25-17-20(12-13-22(25)32(42)45-33-29(39)27(37)28(38)30(44-33)31(40)41)18-26(36)34-23(16-19(2)3)21-10-6-7-11-24(21)35-14-8-5-9-15-35/h6-7,10-13,17,19,23,27-30,33,37-39H,4-5,8-9,14-16,18H2,1-3H3,(H,34,36)(H,40,41)/t23-,27-,28-,29+,30-,33?/m0/s1
Formel vikt 628.2996
Procent renhet >95%