Learn More
Gilson™ REPET-TIPS™ Repeater Pipette Tips
Specifically made for the Gilson™ REPETMAN™ positive-displacement electronic pipette, REPET-TIPS™ cover a range of volumes and are recognized by the REPETMAN™ operating system when placed on the pipette.
3180.00 SEK
Specifications
Filtered | Non-Filtered |
---|---|
Sterility | Sterile |
Color | Clear |
Autoclavable | Not Autoclavable |
Model | REPET-TIP™ |
Product Code | Brand | Volume (Metric) | Graduations | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Graduations | Price | Quantity & Availability | ||||
15344785
|
Gilson™
F164528 |
1 mL | 20 μL |
3180.00 SEK
Pack of 100 |
Estimated Shipment: 17-04-2024
Log in to see stock. |
||||
15354785
|
Gilson™
F164558 |
10 mL | 200 μL |
3180.00 SEK
Pack of 100 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
||||
Description
- Tip-size encoding system of the REPETMAN™ automatically identifies the REPET-TIP™ attached and displays the appropriate volume on the LCD screen
- Made of polypropylene
- Features volume appropriate graduations for precise measurement
Specifications
Non-Filtered | |
Clear | |
REPET-TIP™ | |
66 mm | |
Yes | |
Hinged Rack | |
Repeater Pipette Tips | |
Single-use | |
Yes | |
Polypropylene |
Sterile | |
Not Autoclavable | |
2.5 in | |
ISO 9001, ISO 17025, ISO 8655 | |
Pipetting | |
100 Tips | |
Clear | |
Gilson Repetman | |
REPET-TipS |