Learn More
Gilson™ REPETMAN™ Non-Sterile Tips
These replacement tips are for use with the REPETMAN™ motorized pipette controller.
1645.00 SEK
Specifications
Description | Pipette tips |
---|---|
Sterility | Non-sterile |
For Use With (Application) | Pipetting |
Tip Style | Positive Displacement |
Quantity | 100/pk |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15324785
|
Gilson™
F164527 |
1.0 mL |
1645.00 SEK
Pack of 100 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
15334785
|
Gilson™
F164557 |
10 mL |
1645.00 SEK
Pack of 100 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
Description
- Non-sterile
- Available in 1 and 10mL volumes
- Aids in positive displacement pipetting
- Provided in packs of 100
Specifications
Pipette tips | |
Pipetting | |
100/pk | |
Repetman |
Non-sterile | |
Positive Displacement | |
Repetman |