Learn More
Fisherbrand™ Replacement Burner Head
Guarantees a stable and regular flame.
Brand: Fisherbrand™
Additional Details : Weight : 0.80000kg
Description
- For use with Meker burners
- Grill pattern
- Manufactured from nickel-plated steel
Specifications
Burner Head | |
Meker Burner | |
Nickel-plated Steel | |
Stable and Regular Flame |