Beskrivning
- Compatible with all QuickConnect™ frames
- Multiple lengths available
- Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments.
- Allows investment in one handle with various interchangeable heads required for specific applications.
- Handles are available in several lengths and styles.
- Telescoping handles are available in lightweight carbon fiber, anodised aluminum, durable stainless steel, or fiberglass.
- Choose from a variety of lengths to suit different cleaning needs.
Specifikationer
Specifikationer
| Produkttyp | Cleanroom Mop Handle |
| Sterilitet | Non-sterile |
| Autoklaveringsbar | Autoclavable |
| Inkluderar | 5 Collar Pole |
| Färgkodad | No |
| Material | Carbon Fiber |