Learn More
Labconco™ Replacement Exhaust Filter
Removes oil mist and odor from pump exhaust. Labconco™ Replacement Exhaust Filter includes filter assembly, one oil mist filter cartridge and one odor filter cartridge.
Brand: Labconco™ 1473400
Additional Details : Weight : 2.27000kg
Description
- For General Purpose Vacuum Pumps 1472100, 7739400, 7739402, 1467700, 7739401 and 7739403
- Removes oil mist and odor from pump exhaust
- Ducting to outside not required
- Includes filter assembly, one oil mist filter cartridge and one odor filter cartridge
- Oil mist filter cartridge life is approximately one year and odor filter cartridge life is approximately six months at 1.33 x 10-2 mBar (10 microns) vacuum
Specifications
Rotary Vane Vacuum Pumps | |
Pump Exhaust Filter |