Learn More
Labconco™ Replacement Exhaust Filter
Removes oil mist and odor from pump exhaust. Labconco™ Replacement Exhaust Filter includes filter assembly, one oil mist filter cartridge and one odor filter cartridge.
Brand:  Labconco™ 1473400
Additional Details : Weight : 2.27000kg
Product Code. 15132995
  • 8050.00 SEK / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
  • For General Purpose Vacuum Pumps 1472100, 7739400, 7739402, 1467700, 7739401 and 7739403
  • Removes oil mist and odor from pump exhaust
  • Ducting to outside not required
  • Includes filter assembly, one oil mist filter cartridge and one odor filter cartridge
  • Oil mist filter cartridge life is approximately one year and odor filter cartridge life is approximately six months at 1.33 x 10-2 mBar (10 microns) vacuum
Specifications
Specifications
Rotary Vane Vacuum Pumps
Pump Exhaust Filter