Learn More
Klever Replacement Head (Wider Cut Head)
Product Code. 17799007
Change view
Click to view available options
Quantity:
12/Pk.
Unit Size:
Pack of 240
Product Code. Quantity Unit Size
17799007 12/Pk. Pack of 240
1 options
Product Code. 17799007 Supplier Klever Supplier No. KCJXH30X
  • ONLINE EXCLUSIVE PRICE
    5303.70 SEK
    4229.26 SEK / Paket med 240
    Save 1074.44 SEK
    20% Av
Beräknad leverans: 03-12-2026
Green Replacement Head (wider cut head) for use with Klever X-Change Safety Cutters.
  • Recessed blade reduces cut injuries and damaged goods.
  • Cuts boxes, film, tape, and thousands more applications.
  • Ergonomic soft touch handle reduces hand fatigue and stress.
  • Innovative metal tape splitter
  • Highest safety level
  • Also available in replaceable narrow and multipurpose wide heads
  • Our antimicrobial Kleen™ safety cutters utilize an antimicrobial material that inhibits the growth of bacteria that may affect this product.
  • The antimicrobial properties do not protect users or others against bacteria, viruses, germs, or other disease organisms.
  • Clean and wash this product thoroughly per your industry's guidelines.
  • The active antimicrobial component is part of the formulation, so it won't wash off or wear away. This is not a coating or treatment; it's an integral ingredient.
Specifications
Blade Material Carbon Steel
Product Type Replacement Head
For Use With (Equipment) Kleen™ XChange KCJ-XC-30X
Disposable Reusable
Sterility Non-sterile
Quantity 12/Pk.