Learn More
Trajan Scientific and Medical™ Replacement Needle for NanoVolume Syringes
Range of quality needles to fit a SGE syringe in a range of lengths and tip styles.
Brand: Trajan Scientific and Medical™ 035058
Additional Details : Weight : 0.80000kg
Description
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
23 G | |
0.63 mm | |
Bevel |
0.365 mm | |
5 μL |