Learn More
Trajan Scientific and Medical™ Replacement Plunger for XCHANGE Syringes
Replacement plungers for gas tight syringes.
Brand:  Trajan Scientific and Medical™ 2930480
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15239283
  • 690.00 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
23g
0.63mm
56mm
0.15mm
50μL
Cone
Videos