Learn More
Trajan Scientific and Medical™ Replacement Plunger for XCHANGE Syringes
Replacement plungers for gas tight syringes.
Brand:  Trajan Scientific and Medical™ 2930480
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15239283
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
23 G
0.15 mm
50μL
Cone
0.15mm
0.63mm
56mm
Videos