Learn More
Trajan Scientific and Medical™ Replacement Plungers for CTC Analytics Syringes
Replacement plungers for gas tight syringes.
Brand: Trajan Scientific and Medical™ 031841
Additional Details : Weight : 0.01000kg
Description
Designed to match the specific syringe barrel to maintain a gas tight sealSpecifications
Replacement plunger | |
Fixed Needle | |
CTC Head Space Syringe |
0.47/0.63 mm | |
1 mL |