Learn More
Trajan Scientific and Medical™ Replacement Plungers for CTC Analytics Syringes
Replacement plungers for gas tight syringes.
Brand:  Trajan Scientific and Medical™ 031841
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15298553
  • 1355.00 SEK / Each
Estimated Shipment: 26-07-2024
to see stock.
Description
Description
Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
Replacement plunger
Fixed Needle
CTC Head Space Syringe
0.47/0.63 mm
1 mL