Learn More
Trajan Scientific and Medical™ Replacement Plungers for Fixed Luer-Lock Syringes
Replacement plungers for gas tight syringes.
Brand:  Trajan Scientific and Medical™ 031874
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15218573
  • 1045.00 SEK / Each
Estimated Shipment: 20-06-2024
to see stock.
Description
Description
Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
60 mm
25 mL
LC Priming Pump Syringe