Learn More
Trajan Scientific and Medical™ Replacement Plungers for Fixed Luer-Lock Syringes
Replacement plungers for gas tight syringes.
Brand:  Trajan Scientific and Medical™ 031852
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15248563
  • 785.00 SEK / Each
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
Replacement Plungers
0.17 mm
2.5 mL
Luer Lock Syringe
22 G
0.70 mm
LC