Learn More
Trajan Scientific and Medical™ Replacement Plungers for Hamilton C and CX Syringes
Replacement plungers for gas tight syringes.
Brand:  Trajan Scientific and Medical™ 031864
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15298563
  • 785.00 SEK / Each
Estimated Shipment: 04-07-2024
to see stock.
Description
Description
Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
Replacement Plungers
60 mm
Hamilton C and CX Syringes
GT Plunger and 1/4-28 UNF Termination
10 mL