missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Replacement Plungers for Hamilton C and CX Syringes

Replacement plungers for gas tight syringes.

Brand:  Trajan Scientific and Medical™ 031854

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15258563

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance


Description

Description

Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications

Specifications

±1%
11mm
Hamilton C and CX Syringes
60mm
PTFE
Replacement Plungers
2.5mL
GT Plunger and 1/4-28 UNF Termination
Borosilicate Glass
Videos