Learn More
Trajan Scientific and Medical™ Replacement Plungers for Waters WISP Syringes
Replacement plungers for gas tight syringes.
Brand:  Trajan Scientific and Medical™ 031819
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15238543
  • 449.00 SEK / Each
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
Replacement plunger
25μL
60mm
1/4-28 UNF screw thread
Videos