Learn More
Trajan Scientific and Medical™ Replacement Plungers for Waters WISP Syringes
Replacement plungers for gas tight syringes.
Brand:  Trajan Scientific and Medical™ 031819
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15238543
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
Description
Description
Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
Replacement plunger
25μL
60mm
1/4-28 UNF screw thread
Videos