Learn More
Thermo Scientific™ Replacement Needles for LC Syringes
Thermo Scientific™ Replacement Needles for LC Syringes are available for syringes with removable needles.
Brand:  Thermo Scientific™ 365RNL25
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10079142
  • 329.46 SEK / Pack of 5
Estimated Shipment: 05-01-2022
Description
Description
Thermo Scientific™ Replacement Needles for LC Syringes are available for syringes with removable needles.
Specifications
Specifications
365RNL25
2 in.
Syringe Needle
LC Autosampler Syringe
22 G
25 to 500 µL
50.8 mm
Removable