Läs mer
Tocris Bioscience™ RepSox
Beskrivning
RepSox is an ancillary Material Grade version of product RepSox (Cat. No. 3742). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.
Find out more about our Ancillary Material Grade product range.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2-[3-(6-Methylpyridin-2-yl)-1 H-pyrazol-4-yl]-1,5-naphthyridine |
| CAS | 446859-33-2 |
| Kvantitet | 10 mg |
| Mål | TGF-beta Receptor Inhibitors |
| Molekylformel | C17H13N5 |
| Renhet | 0.99 |