Läs mer
Reserpic Acid Methyl Ester, TRC
Produktkod. 30412564
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412564 50 mg
30416125 5 mg
30420452 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30412564 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR14459050MG
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2901-66-8
Molekylformel C23H30N2O5
Molekylvikt (g/mol) 414.49
Synonym Methyl reserpate, Methyl reserpinolate, NSC 81464, (3β,16β,17α,18β,20α)-18-Hydroxy-11,17-dimethoxy-yohimban-16-carboxylic Acid Methyl Ester
IUPAC-namn methyl (1R,15S,17R,18R,19S,20S)-17-hydroxy-6,18-dimethoxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
LEDER COC1=CC2=C(C=C1)C3=C([C@H]4C[C@H]5[C@H](C[C@H]([C@@H]([C@H]5C(=O)OC)OC)O)CN4CC3)N2
Specifikationer
Kemiskt namn eller material Reserpic Acid Methyl Ester
Kvantitet 50 mg
InChI formel InChI=1S/C23H30N2O5/c1-28-13-4-5-14-15-6-7-25-11-12-8-19(26)22(29-2)20(23(27)30-3)16(12)10-18(25)21(15)24-17(14)9-13/h4-5,9,12,16,18-20,22,24,26H,6-8,10-11H2,1-3H3/t12-,16+,18-,19-,20+,22+/m1/s1
Formel vikt 414.2155