Läs mer
Resibufogenin, TRC
Produktkod. 30421309
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30421309 25 mg
1 options
Produktkod. 30421309 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR14466025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Resibufogenin
CAS 465-39-4
Molekylformel C24 H32 O4
Kvantitet 25 mg
Synonym Bufa-20,22-dienolide','14,15-epoxy-3-hydroxy-','(3β,5β,15β)-, 5β-Bufa-20,22-dienolide','14,15β-epoxy-3β-hydroxy- (6CI,7CI,8CI), (3β,5β,15β)-14,15-Epoxy-3-hydroxybufa-20,22-dienolide, 14,15β-Epoxy-3β-hydroxy-5β-bufa-20,22-dienolide, 3β-Hydroxy-14,15β-epoxy-5β-bufa-20,22-dienolide, Bufogenin, NSC 90783, Recibufogenin, Resi, Resibufogenin, Resinobufagin, Respigon
InChI formel InChI=1S/C24H32O4/c1-22-9-7-16(25)11-15(22)4-5-18-17(22)8-10-23(2)19(12-20-24(18,23)28-20)14-3-6-21(26)27-13-14/h3,6,13,15-20,25H,4-5,7-12H2,1-2H3/t15-,16+,17+,18-,19-,20-,22+,23-,24-/m1/s1
LEDER C[C@]12CC[C@H](O)C[C@H]1CC[C@@H]3[C@@H]2CC[C@]4(C)[C@H](C[C@H]5O[C@@]345)C6=COC(=O)C=C6
Molekylvikt (g/mol) 384.51
Formel vikt 384.2301