Läs mer
Resmetirom, MedChemExpress
Produktkod. 16487747
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16487747 5 mg
16445776 100 mg
16425028 10 mg
16411267 50 mg
16410156 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16487747 Leverantör Medchem Express Leverantörsnummer HY12216
MedChemExpress Resmetirom (MGL-3196) is a highly selective thyroid hormone receptor β (THR-β) agonist with an EC50 value of 0.21 μM.
Resmetirom (MGL-3196) is a highly selective thyroid hormone receptor β (THR-β) agonist with an EC50 value of 0.21 μM.
Specifikationer
Kemiskt namn eller material Resmetirom
CAS 920509-32-6
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C17H12Cl2N6O4
Kvantitet 5 mg
Synonym MGL-3196 VIA-3196
Löslighetsinformation DMSO : ≥ 31 mg/mL (71.23 mM)
LEDER N#CC1=NN(C2=CC(Cl)=C(OC(C=C3C(C)C)=NNC3=O)C(Cl)=C2)C(NC1=O)=O
Molekylvikt (g/mol) 435.22
Formel vikt 435.22
Procent renhet 98.02%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation