Läs mer
Resorcinol-13C6, TRC
Produktkod. 30416703
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30416703 50 mg
30416702 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30416703 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR14470350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 953390-31-3
Molekylformel 13C6 H6 O2
Molekylvikt (g/mol) 116.07
Synonym 1,3-Benzenediol-1,2,3,4,5,6-13C6, 1,3-Dihydroxybenzene-13C6, 3-Hydroxyphenol-13C6, m-Hydroquinone-13C6, m-Hydroxyphenol-13C6, m-Phenylenediol-13C6, m-Benzenediol-13C6, m-Dihydroxybenzene-13C6
IUPAC-namn (1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene-1,3-diol
LEDER O[13c]1[13cH][13cH][13cH][13c](O)[13cH]1
Specifikationer
Kemiskt namn eller material Resorcinol-13C6
Kvantitet 50 mg
InChI formel InChI=1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 116.0569