Learn More
Moldex™ Respirator FFP1S Disposable
Suited to most industrial applications this general purpose mask is strong and durable
Brand: Moldex™ 238001
Additional Details : Weight : 0.38250kg
Description
Moldex Smart Series FFP1 NR D non-valved dust mask. Suited to most industrial applications this general purpose mask is strong and durable. The ActiveForm™ seal automatically adjusts to different face shapes and movements to give a secure seal.Specifications
Single-use | |
Respirator FFP1S Disposable | |
20/Pk. |