Learn More
Moldex™ Respirator individually packed cup type, foldable, FFP2, unvalved Moldex
Brand:  Moldex™ 247001
Additional Details : Weight : 0.21432kg
Product Code. 12302329
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
Description
Moldex Smart Pocket FFP2 NR D non-valved dust mask.
This foldable pocket mask has all the features of a traditional Moldex cup mask. Ideally suited to lighter industries, where the mask is carried between use or industries with hygiene implications.