Läs mer
Resveratrol-13C6, TRC
Produktkod. 30135675
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135675 10 mg
30003689 5 mg
30005199 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135675 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR15000210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1185247-70-4
Molekylformel 13C6 C8 H12 O3
Molekylvikt (g/mol) 234.2
Synonym Resveratrol-13C6, Resveratrol-(4-hydroxyphenyl-13C6), 5-[2-(4-Hydroxyphenyl-13C6)ethenyl]-1,3-benzenediol, (EZ)-Resveratrol-13C6 (4-hydroxyphenyl-13C6)
IUPAC-namn 5-[2-(4-hydroxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)ethenyl]benzene-1,3-diol
LEDER O[13c]1[13cH][13cH][13c]([13cH][13cH]1)C=Cc2cc(O)cc(O)c2
Specifikationer
Kemiskt namn eller material (EZ)-Resveratrol 4-hydroxyphenyl-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C14H12O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h1-9,15-17H/i3+1,4+1,5+1,6+1,10+1,12+1
Formel vikt 234.0988
Procent renhet >95%