Läs mer
Retapamulin, TRC
Produktkod. 30422922
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30422922 500 mg
1 options
Produktkod. 30422922 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR279008500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Retapamulin
CAS 224452-66-8
Molekylformel C30 H47 N O4 S
Kvantitet 500 mg
Synonym Acetic acid','2-[[(3-exo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl]thio]-','(3aS,4R,5S,6S,8R,9R,9aR,10R)-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester, Acetic acid','[[(3-exo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl]thio]-','(3aS,4R,5S,6S,8R,9R,9aR,10R)-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester (9CI), Altabax, Altargo, Mutilin 14-(exo-8-methyl-8-azabicyclo[3.2.1]oct-3-ylsulfanyl)acetate, Retapamulin, SB 275833, SB-275833, SB-275833, GSK 527144A, GSK 1091913A, SB-278968, GSK 395946A, GSK 990983A, GSK 1058377A
InChI formel InChI=1S/C30H47NO4S/c1-7-28(4)16-24(35-25(33)17-36-22-14-20-8-9-21(15-22)31(20)6)29(5)18(2)10-12-30(19(3)27(28)34)13-11-23(32)26(29)30/h7,18-22,24,26-27,34H,1,8-17H2,2-6H3/t18-,19+,20-,21+,22-,24-,26+,27+,28-,29?,30?/m1/s1
LEDER C[C@@H]1CCC23CCC(=O)[C@H]2C1(C)[C@@H](C[C@@](C)(C=C)[C@@H](O)[C@@H]3C)OC(=O)CS[C@@H]4C[C@H]5CC[C@@H](C4)N5C
Molekylvikt (g/mol) 517.76
Formel vikt 517.3226