missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reticulocalbin 3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27497-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Calcipressin 3 inhibits calcineurin dependent transcriptional responses by binding to the catalytic domain of calcineurin A, and may play a role during central nervous system development. Highest expression occurs in heart, skeletal muscle kidney, liver and peripheral blood leukocytes.Specifications
Reticulocalbin 3 | |
Polyclonal | |
Unconjugated | |
RCN3 | |
RCN3, Reticulocalbin 3, RLP49 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57333 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96D15 | |
RCN3 | |
Reticulocalbin 3 Fusion Protein Ag26569 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |