Läs mer
Retin-15-yl beta-D-Glucopyranosiduronic Acid(>85%), TRC
Produktkod. 30411378
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30411378 10 mg
1 options
Produktkod. 30411378 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR23984010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Retin-15-yl beta-D-Glucopyranosiduronic Acid
CAS 16639-19-3
Molekylformel C26H38O7
Kvantitet 10 mg
Synonym β-D-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenyl Glucopyranosiduronic Acid, (all-E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenyl β-D-Glucopyranosiduronic Acid, Retin-15-yl β-D-Glucopyranosiduronic Acid
InChI formel InChI=1S/C26H38O7/c1-16(11-12-19-18(3)10-7-14-26(19,4)5)8-6-9-17(2)13-15-32-25-22(29)20(27)21(28)23(33-25)24(30)31/h6,8-9,11-13,20-23,25,27-29H,7,10,14-15H2,1-5H3,(H,30,31)/b9-6+,12-11+,16-8+,17-13+/t20-,21-,22+,23-,25+/m0/s1
LEDER CC1(C)C(/C=C/C(C)=C/C=C/C(C)=C/CO[C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O2)=C(C)CCC1
IUPAC-namn (2S,3S,4S,5R,6R)-6-(((2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid
Molekylvikt (g/mol) 462.58
Formel vikt 462.26