missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Retinal S antigen Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Retinal S antigen |
---|---|
Concentration | 0.52 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16822385
|
Proteintech
23546-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16812385
|
Proteintech
23546-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
S-antigen, also identified as arrestin, is a protein involved in the regulation of phototransduction.Specifications
Retinal S antigen | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P10523, P20443 | |
20215, 6295 | |
Retinal S antigen Fusion Protein Ag20318 | |
Primary | |
-20°C | |
SAG |
0.52 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
48 kDa protein, Retinal S antigen, Rod photoreceptor arrestin, S AG, S arrestin, SAG | |
SAG | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |