Läs mer
Retinyl Propionate (Stabilized with BHT) 90%, TRC
Produktkod. 30137698
Change view
Kvantitet:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137698 5 g
30006855 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137698 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR2755505G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7069-42-3
Molekylformel C23 H34 O2
Molekylvikt (g/mol) 342.51
Synonym Retinol, 15-propanoate, Retinol propionate (7CI), Retinol, propanoate (9CI), Retinol, propionate, all-trans- (8CI), Retinyl propionate, Vitamin A propionate
IUPAC-namn [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] propanoate
LEDER CCC(=O)OC\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
Specifikationer
Kemiskt namn eller material Retinyl Propionate
Kvantitet 5 g
InChI formel InChI=1S/C23H34O2/c1-7-22(24)25-17-15-19(3)11-8-10-18(2)13-14-21-20(4)12-9-16-23(21,5)6/h8,10-11,13-15H,7,9,12,16-17H2,1-6H3/b11-8+,14-13+,18-10+,19-15+
Formel vikt 342.2559