Läs mer
Retinyl Retinoate, TRC
Produktkod. 30427552
Change view
Kvantitet:
10 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427552 25 mg
30414120 250 mg
30409366 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427552 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR27555525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15498-86-9
Molekylformel C40H56O2
Molekylvikt (g/mol) 568.89
Synonym Retinoic Acid Retin-15-yl Ester, Retinoic Acid Ester with Retinol
IUPAC-namn (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraen-1-yl (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoate
LEDER O=C(OC/C=C(C)/C=C/C=C(C)/C=C/C1=C(C)CCCC1(C)C)/C=C(C)/C=C/C=C(C)/C=C/C2=C(C)CCCC2(C)C
Specifikationer
Kemiskt namn eller material Retinyl Retinoate
Kvantitet 25 mg
InChI formel InChI=1S/C40H56O2/c1-30(21-23-36-34(5)19-13-26-39(36,7)8)15-11-17-32(3)25-28-42-38(41)29-33(4)18-12-16-31(2)22-24-37-35(6)20-14-27-40(37,9)10/h11-12,15-18,21-25,29H,13-14,19-20,26-28H2,1-10H3/b17-11+,18-12+,23-21+,24-22+,30-15+,31-16+,32-25+,33-29+
Formel vikt 568.43